Stampa…

Bis(4-biphenylyl)amine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B2429-1G B2429-5G
TCIAB2429-1GEA 21 EUR
TCIAB2429-1G TCIAB2429-5G
Bis(4-biphenylyl)amine ≥98.0% (by GC)
Bis(4-biphenylyl)amine
Formula: C₂₄H₁₉N
MDL Number: MFCD08276279
CAS Number: 102113-98-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 207.0 to 211.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR