sec-Butyl ethyl ether ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
B2399-25G
B2399-250G
TCIAB2399-25GEA
100
EUR
TCIAB2399-25G
TCIAB2399-250G
sec-Butyl ethyl ether ≥97.0% (by GC)
sec-Butyl ethyl ether
Formula:
C₆H₁₄O Punto di ebollizione: 82°C |
MDL Number:
MFCD00053773 CAS Number: 2679-87-0 EINECS: 220-234-9 UN: 1179 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...