Bis(4-tert-butylphenyl)iodonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Bis(4-(tert-butyl)phenyl)iodonium hexafluorophosphate(V)
B2380-1G
B2380-5G
TCIAB2380-1GEA
55
EUR
TCIAB2380-1G
TCIAB2380-5G
Bis(4-tert-butylphenyl)iodonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Bis(4-tert-butylphenyl)iodonium hexafluorophosphate
Formula:
C₂₀H₂₆F₆IP Peso molecolare: 538,30 g/mol |
MDL Number:
MFCD06797070 CAS Number: 61358-25-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...