Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
B2358-1G
B2358-5G
TCIAB2358-1GEA
80
EUR
TCIAB2358-1G
TCIAB2358-5G
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥98.0% (by HPLC)
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate
Formula:
C₁₆H₂₂BrF₆N₂P Temperatura di stoccaggio: Refrigerata |
CAS Number:
188944-77-6 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...