Stampa…

4-(4-Butylphenylazo)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Butyl-4'-hydroxyazobenzene

B2357-25G B2357-5G
TCIAB2357-25GEA 211 EUR
TCIAB2357-25G TCIAB2357-5G
4-(4-Butylphenylazo)phenol ≥98.0% (by GC)
4-(4-Butylphenylazo)phenol
Formula: C₁₆H₁₈N₂O
Peso molecolare: 254,33 g/mol
Punto di fusione: 80°C
MDL Number: MFCD00430795
CAS Number: 2496-21-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 82.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR