Stampa…

4-tert-Butylsulphonylcalix[4]arene ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Tetra-tert-butyl(tetrahydroxy)tetrasulphonylcalix[4]arene, 15,35,55,75-Tetra-tert-butyl-12,32,52,72-tetrahydroxy-2,4,6,8-tetrathia-1,3,5,7(1,3)-tetrabenzenacyclooctaphane 2,2,4,4,6,6,8,8-octaoxide

B2312-1G B2312-5G
TCIAB2312-1GEA 145 EUR
TCIAB2312-1G TCIAB2312-5G
4-tert-Butylsulphonylcalix[4]arene ≥98.0% (by HPLC)
4-tert-Butylsulphonylcalix[4]arene
Formula: C₄₀H₄₈O₁₂S₄
Peso molecolare: 849,05 g/mol
MDL Number: MFCD06797060
CAS Number: 204190-49-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR