Stampa…

4-tert-Butylthiacalix[4]arene ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetra-tert-butyl(tetrahydroxy)tetrathiacalix[4]arene

B2296-1G B2296-5G
TCIAB2296-1GEA 32 EUR
TCIAB2296-1G TCIAB2296-5G
4-tert-Butylthiacalix[4]arene ≥98.0% (by HPLC)
4-tert-Butylthiacalix[4]arene
Formula: C₄₀H₄₈O₄S₄
Peso molecolare: 721,06 g/mol
MDL Number: MFCD04038412
CAS Number: 182496-55-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Lambda max. 294.0 to 297.0 nm (THF)
Absorbance (E1%1cm) min. 280 (THF, 294.0 to 297.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR