Stampa…

1-[Bis(trifluoromethanesulphonyl)methyl]-2,3,4,5,6-pentafluorobenzene ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: 2,3,4,5,6-Pentafluorophenylbis(trifluoromethanesulphonyl)methane, alpha,alpha-Bis(trifluoromethanesulphonyl)-2,3,4,5,6-pentafluorotoluene

B2291-1G B2291-100MG
TCIAB2291-1GEA 484 EUR
TCIAB2291-1G TCIAB2291-100MG
1-[Bis(trifluoromethanesulphonyl)methyl]-2,3,4,5,6-pentafluorobenzene ≥98.0% (by titrimetric analysis)
1-[Bis(trifluoromethanesulphonyl)methyl]-2,3,4,5,6-pentafluorobenzene
Formula: C₉HF₁₁O₄S₂
MDL Number: MFCD04117910
CAS Number: 405074-81-9
UN: 3261
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR