Stampa…

1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥97.0% (by GC)

Fornitore: TCI EUROPE

B2287-100MG B2287-1G
TCIAB2287-100MGEA 86 EUR
TCIAB2287-100MG TCIAB2287-1G
1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥97.0% (by GC)
1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene
Formula: C₂₃H₁₄F₆S₂
Peso molecolare: 468,48 g/mol
MDL Number: MFCD06656279
CAS Number: 137814-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a903fb9e006e684937102f165b8423f22667fb1&vid=1a903fb9e006e684937102f165b8423f22667fb1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 157.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR