Stampa…

Ethyl-1-benzylpyrrole-3-carboxylate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester

B2284-1G B2284-5G
TCIAB2284-1GEA 76 EUR
TCIAB2284-1G TCIAB2284-5G
Ethyl-1-benzylpyrrole-3-carboxylate ≥95.0% (by GC)
Ethyl-1-benzylpyrrole-3-carboxylate
Formula: C₁₄H₁₅NO₂
Peso molecolare: 229,28 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06797056
CAS Number: 128259-47-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR