Stampa…

3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B2247-1G B2247-5G
TCIAB2247-1GEA 146 EUR
TCIAB2247-1G TCIAB2247-5G
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide ≥97.0% (by HPLC, titration analysis)
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide
Formula: C₂₅H₂₇BrO₆
Peso molecolare: 503,39 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD04038410
CAS Number: 176650-93-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR