Stampa…

(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Boc-Phe(4-NO2)-OH, Boc-Phe(4-NO₂)-OH, N-Boc-4-nitro-L-phenylalanine, Boc-p-nitro-Phe-OH

B2232-1G B2232-5G
TCIAB2232-1GEA 28 EUR
TCIAB2232-1G TCIAB2232-5G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid ≥98.0% (by titrimetric analysis)
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid
Formula: C₁₄H₁₈N₂O₆
Peso molecolare: 310,3 g/mol
MDL Number: MFCD00038128
CAS Number: 33305-77-0
EINECS: 251-450-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +5.0 to +10.0 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR