Stampa…

2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (R,R)-Ph-pybox, (+)-2,6-Bis[(4R)-4-phenyl-2-oxazolin-2-yl]pyridine

B2219-1G B2219-250MG
TCIAB2219-1GEA 95 EUR
TCIAB2219-1G TCIAB2219-250MG
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98.0% (by HPLC, titration analysis)
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine
Formula: C₂₃H₁₉N₃O₂
Peso molecolare: 369,42 g/mol
MDL Number: MFCD01863585
CAS Number: 128249-70-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR