Stampa…

Boc-b-Alanine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine

B2176-25G B2176-5G
TCIAB2176-25GEA 53 EUR
TCIAB2176-25G TCIAB2176-5G
Boc-b-Alanine ≥98.0% (by HPLC, titration analysis)
Boc-b-Alanine
Formula: C₈H₁₅NO₄
Peso molecolare: 189,2 g/mol
MDL Number: MFCD00037291
CAS Number: 3303-84-2
EINECS: 221-979-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR