Stampa…

2-Bromophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Bromophenylhydrazine hydrochloride, 1-(2-Bromophenyl)hydrazine hydrochloride

B2147-5G B2147-25G
TCIAB2147-5GEA 57 EUR
TCIAB2147-5G TCIAB2147-25G
2-Bromophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
2-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
MDL Number: MFCD00012926
CAS Number: 50709-33-6
EINECS: 256-728-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 % (calcd.on anh.substance)
Water (Vaporization methoed) max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR