Stampa…

(R)-(+)-3-Benzyloxy-1,2-propanediol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (R)-(+)-Glycerol alpha-Benzyl Ether, (R)-3-(Benzyloxy)propane-1,2-diol

B2141-1G B2141-100MG
TCIAB2141-1GEA 100 EUR
TCIAB2141-1G TCIAB2141-100MG
(R)-(+)-3-Benzyloxy-1,2-propanediol ≥98.0% (by GC)
(R)-(+)-3-Benzyloxy-1,2-propanediol
Formula: C₁₀H₁₄O₃
Peso molecolare: 182,22 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00067260
CAS Number: 56552-80-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR