Stampa…

Methyl (S)-(-)-3-bromoisobutyrate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-(-)-3-Bromoisobutyric Acid Methyl Ester, Methyl (S)-(-)-3-Bromo-2-methylpropionate, (S)-(-)-3-Bromo-2-methylpropionic Acid Methyl Ester, Methyl (S)-3-bromo-2-methylpropanoate

B2140-1G B2140-5G
TCIAB2140-1GEA 58 EUR
TCIAB2140-1G TCIAB2140-5G
Methyl (S)-(-)-3-bromoisobutyrate ≥97.0% (by GC)
Methyl (S)-(-)-3-bromoisobutyrate
Formula: C₅H₉BrO₂
Peso molecolare: 181,03 g/mol
Flash Pt: 73°C
MDL Number: MFCD00064487
CAS Number: 98190-85-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Optical purity (GC) min. 99.0 ee%
Specific rotation [a]20/D -13.0 to -19.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR