4-[(tert-Butoxycarbonylamino)methyl]pyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(Boc-aminomethyl)pyridine
B2126-25G
B2126-5G
TCIAB2126-25GEA
367
EUR
TCIAB2126-25G
TCIAB2126-5G
4-[(tert-Butoxycarbonylamino)methyl]pyridine ≥98.0% (by GC, titration analysis)
4-[(tert-Butoxycarbonylamino)methyl]pyridine
Formula:
C₁₁H₁₆N₂O₂ Peso molecolare: 208,26 g/mol |
MDL Number:
MFCD01321573 CAS Number: 111080-65-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...