9,10-Bis(3,5-dihydroxyphenyl)anthracene ≥96.0% (by HPLC)
Fornitore: TCI
Formula:
C₂₆H₁₈O₄ |
MDL Number:
MFCD03427350 CAS Number: 153715-08-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...