Stampa…

(1R,2S)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: (1R,2S)-N-Benzyl-N-(mesitylenesulphonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulphonyl)amino]-1-phenyl-1-propanol

B2103-1G B2103-5G
TCIAB2103-1GEA 93 EUR
TCIAB2103-1G TCIAB2103-5G
(1R,2S)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol ≥98.0% (by HPLC, total nitrogen)
(1R,2S)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenyl-1-propanol
Formula: C₂₅H₂₉NO₃S
Peso molecolare: 423,57 g/mol
MDL Number: MFCD02093446
CAS Number: 187324-63-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 130.0 to 134.0 deg-C
Specific rotation [a]20/D +48.0 to +52.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR