Trihexyl borate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Boric Acid Trihexyl Ester
B2094-25G
B2094-5G
TCIAB2094-25GEA
78
EUR
TCIAB2094-25G
TCIAB2094-5G
Trihexyl borate ≥98.0% (by titrimetric analysis)
Trihexyl borate
Formula:
C₁₈H₃₉BO₃ Peso molecolare: 314,32 g/mol Punto di ebollizione: 311°C Flash Pt: 149°C |
MDL Number:
MFCD00027295 CAS Number: 5337-36-0 EINECS: 226-262-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...