Stampa…

Bis[2-(2-benzothiazolyl)phenolato]zinc(II) ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

B2077-1G B2077-5G
TCIAB2077-1GEA 47 EUR
TCIAB2077-1G TCIAB2077-5G
Bis[2-(2-benzothiazolyl)phenolato]zinc(II) ≥98.0% (by titrimetric analysis)
Bis[2-(2-benzothiazolyl)phenolato]zinc(II)
Formula: C₂₆H₁₆N₂O₂S₂Zn
MDL Number: MFCD03844766
CAS Number: 58280-31-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR