Stampa…

1,1-Bis(4-aminophenyl)cyclohexane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-Cyclohexylidenedianiline

B2076-25G B2076-5G
TCIAB2076-25GEA 448 EUR
TCIAB2076-25G TCIAB2076-5G
1,1-Bis(4-aminophenyl)cyclohexane ≥98.0% (by GC)
1,1-Bis(4-aminophenyl)cyclohexane
Formula: C₁₈H₂₂N₂
Peso molecolare: 266,39 g/mol
MDL Number: MFCD00035700
CAS Number: 3282-99-3
EINECS: 221-926-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR