Stampa…

2,2-Bis(hydroxymethyl)butyric acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2,2-Dimethylolbutyric Acid

B2038-25G B2038-500G
TCIAB2038-25GEA 16 EUR
TCIAB2038-25G TCIAB2038-500G
2,2-Bis(hydroxymethyl)butyric acid ≥97.0% (by titrimetric analysis)
2,2-Bis(hydroxymethyl)butyric acid
Formula: C₆H₁₂O₄
Peso molecolare: 148,16 g/mol
MDL Number: MFCD00190088
CAS Number: 10097-02-6
EINECS: 424-090-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 110.0 to 118.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR