Stampa…

[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,4-Butylenebis(diphenylphosphine)palladium dichloride, Dichloro[1,4-bis(diphenylphosphino)butane]palladium(II)

B2031-1G B2031-5G
TCIAB2031-1GEA 110 EUR
TCIAB2031-1G TCIAB2031-5G
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride ≥96.0% (by titrimetric analysis)
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride
Formula: C₂₈H₂₈Cl₂P₂Pd
Peso molecolare: 603,80 g/mol
MDL Number: MFCD02093437
CAS Number: 29964-62-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR