2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Tetrabromobisphenol A Bis(2,3-dibromopropyl) Ether, 2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane(Granular)
B2022-25G
B2022-500G
TCIAB2022-25GEA
17
EUR
TCIAB2022-25G
TCIAB2022-500G
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane ≥95.0% (by titrimetric analysis)
2,2-Bis[3,5-dibromo-4-(2,3-dibromopropoxy)phenyl]propane
Formula:
C₂₁H₂₀Br₈O₂ Peso molecolare: 943,62 g/mol |
MDL Number:
MFCD00017887 CAS Number: 21850-44-2 EINECS: 244-617-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 94.0 area% |
Purity (Argentmetric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...