2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane ≥99.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Tetrabromobisphenol A Diallyl Ether
B2021-25G
B2021-500G
TCIAB2021-25GEA
16
EUR
TCIAB2021-25G
TCIAB2021-500G
2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane ≥99.0% (by HPLC)
2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane
Formula:
C₂₁H₂₀Br₄O₂ Peso molecolare: 624,01 g/mol |
MDL Number:
MFCD00086863 CAS Number: 25327-89-3 EINECS: 246-850-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Melting point | 118.0 to 122.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...