Stampa…

2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde, 1,4-Benzodioxan-6-carboxaldehyde, 2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde

B2019-1G B2019-5G
TCIAB2019-1GEA 20 EUR
TCIAB2019-1G TCIAB2019-5G
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde ≥98.0% (by GC)
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
Formula: C₉H₈O₃
Peso molecolare: 164,16 g/mol
Punto di ebollizione: 299°C
MDL Number: MFCD00010092
CAS Number: 29668-44-8
EINECS: 249-763-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 53.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR