Stampa…

N-(tert-Butoxycarbonyl)-L-serine-β-lactone ≥98.0% (by total nitrogen basis)

Fornitore: TCI

B1995-1G B1995-5G
TCIAB1995-1GEA 264 EUR
TCIAB1995-1G TCIAB1995-5G
N-(tert-Butoxycarbonyl)-L-serine-β-lactone ≥98.0% (by total nitrogen basis)
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone
Formula: C₈H₁₃NO₄
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD01318414
CAS Number: 98541-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffae7f38107e46ba9a6885c77b792b64b918d36f&vid=ffae7f38107e46ba9a6885c77b792b64b918d36f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -21.0 to -31.0 deg (C=1, CH3CN)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR