(2-Benzothiazolylthio)acetic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[(1,3-Benzothiazol-2-yl)sulphanyl]acetic acid, 2-(Benzo[d]thiazol-2-ylthio)acetic acid
B1941-25G
B1941-500G
TCIAB1941-25GEA
17
EUR
TCIAB1941-25G
TCIAB1941-500G
(2-Benzothiazolylthio)acetic acid ≥98.0% (by HPLC, titration analysis)
(2-Benzothiazolylthio)acetic acid
Formula:
C₉H₇NO₂S₂ Peso molecolare: 225,28 g/mol |
MDL Number:
MFCD00227391 CAS Number: 6295-57-4 EINECS: 228-565-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 156.0 to 160.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...