Benzyl chloromethyl sulphide ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
alpha-(Chloromethylthio)toluene
B1937-25G
B1937-5G
TCIAB1937-25GEA
292
EUR
TCIAB1937-25G
TCIAB1937-5G
Benzyl chloromethyl sulphide ≥96.0% (by GC)
Benzyl chloromethyl sulphide
Formula:
C₈H₉ClS Peso molecolare: 172,67 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD02093436 CAS Number: 3970-13-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...