4-Octadecyloxybiphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Stearyloxybiphenyl, Biphenyl-4-yl Octadecyl Ether, Biphenyl-4-yl Stearyl Ether
B1924-25G
B1924-5G
TCIAB1924-25GEA
125
EUR
TCIAB1924-25G
TCIAB1924-5G
4-Octadecyloxybiphenyl ≥98.0% (by GC)
4-Octadecyloxybiphenyl
Formula:
C₃₀H₄₆O Peso molecolare: 422,70 g/mol |
MDL Number:
MFCD01321145 CAS Number: 376609-78-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...