3-Bromo-1-(triisopropylsilyl)pyrrole ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-bromo-1-[tris(propan-2-yl)silyl]-1H-pyrrole
B1899-1G
B1899-25G
B1899-5G
TCIAB1899-1GEA
63
EUR
TCIAB1899-1G
TCIAB1899-25G
TCIAB1899-5G
3-Bromo-1-(triisopropylsilyl)pyrrole ≥95.0% (by GC)
3-Bromo-1-(triisopropylsilyl)pyrrole
Formula:
C₁₃H₂₄BrNSi Peso molecolare: 302,33 g/mol |
MDL Number:
MFCD01321136 CAS Number: 87630-36-2 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...