Stampa…

Bis(2-ethylhexanoato)hydroxyaluminum ≥96.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

B1895-25G B1895-250G
TCIAB1895-25GEA 17 EUR
TCIAB1895-25G TCIAB1895-250G
Bis(2-ethylhexanoato)hydroxyaluminum ≥96.0% (by titrimetric analysis)
Bis(2-ethylhexanoato)hydroxyaluminum
Formula: C₁₆H₃₁AlO₅
MDL Number: MFCD00041940
CAS Number: 30745-55-2
EINECS: 250-322-2
UN: 1325
ADR: 4.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 96.0 %
Solubility in hot 1mol/L NaOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR