1,3-Bis(tosyloxy)-2,2-dimethylpropane ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
1,3-Bis(p-toluenesulphonyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Ditosylate, Neopentyl Glycol Ditosylate
B1870-25G
B1870-5G
TCIAB1870-25GEA
130
EUR
TCIAB1870-25G
TCIAB1870-5G
1,3-Bis(tosyloxy)-2,2-dimethylpropane ≥98.0% (by GC)
1,3-Bis(tosyloxy)-2,2-dimethylpropane
Formula:
C₁₉H₂₄O₆S₂ Peso molecolare: 412,52 g/mol |
MDL Number:
MFCD00110766 CAS Number: 22308-12-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...