Stampa…

tert-Butyl-N-(Benzyloxy)carbamate ≥98.0% (by total nitrogen basis)

Fornitore: TCI
Attenzione

Synonyms: tert-Butyl benzyloxycarbamate, N-(Benzyloxy)carbamic Acid tert-Butyl Ester

B1799-25G B1799-5G
TCIAB1799-25GEA 186 EUR
TCIAB1799-25G TCIAB1799-5G
tert-Butyl-N-(Benzyloxy)carbamate ≥98.0% (by total nitrogen basis)
tert-Butyl-N-(Benzyloxy)carbamate
Formula: C₁₂H₁₇NO₃
MDL Number: MFCD00191873
CAS Number: 79722-21-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR