Stampa…

1,3-Bis(vinylsulphonyl)-2-propanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

B1745-1G B1745-5G
TCIAB1745-1GEA 89 EUR
TCIAB1745-1G TCIAB1745-5G
1,3-Bis(vinylsulphonyl)-2-propanol ≥98.0% (by GC)
1,3-Bis(vinylsulphonyl)-2-propanol
Formula: C₇H₁₂O₅S₂
Peso molecolare: 240,29 g/mol
MDL Number: MFCD00671498
CAS Number: 67006-32-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR