Stampa…

Diethyl-N-(tert-butoxycarbonyl)phosphoramidate ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Diethyl N-Boc-phosphoramidate, N-Boc-phosphoramidic Acid Diethyl Ester, N-(tert-Butoxycarbonyl)phosphoramidic Acid Diethyl Ester

B1734-5G B1734-1G
TCIAB1734-5GEA 248 EUR
TCIAB1734-5G TCIAB1734-1G
Diethyl-N-(tert-butoxycarbonyl)phosphoramidate ≥98.0% (by HPLC, total nitrogen)
Diethyl-N-(tert-butoxycarbonyl)phosphoramidate
Formula: C₉H₂₀NO₅P
Peso molecolare: 253,23 g/mol
MDL Number: MFCD00015667
CAS Number: 85232-02-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 64.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR