1,3-Bis(4-methoxyphenyl)-1,3-propanedione ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₇H₁₆O₄ |
MDL Number:
MFCD00025817 CAS Number: 18362-51-1 EINECS: 242-233-2 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...