Stampa…

2,2'-Bis(trifluoromethyl)benzidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine, 4,4'-Diamino-2,2'-bis(trifluoromethyl)biphenyl

B1711-25G B1711-5G B1711-1G
TCIAB1711-25GEA 89 EUR
TCIAB1711-25G TCIAB1711-5G TCIAB1711-1G
2,2'-Bis(trifluoromethyl)benzidine ≥98.0% (by GC, titration analysis)
2,2'-Bis(trifluoromethyl)benzidine
Formula: C₁₄H₁₀F₆N₂
Peso molecolare: 320,24 g/mol
MDL Number: MFCD00190155
CAS Number: 341-58-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR