Nε-(tert-butoxycarbonyl)-L-lysine ≥97.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
Nε-Boc-L-lysine, H-Lys(Boc)-OH
Formula:
C₁₁H₂₂N₂O₄ |
MDL Number:
MFCD00037221 CAS Number: 2418-95-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Specific rotation [a]20/D | +16.0 to +19.0 deg (C=1, AcOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...