Stampa…

2,2'-Bis(hydroxymethyl)diphenyl ether ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,2'-Oxydi(benzyl Alcohol)

B1643-1G B1643-5G
TCIAB1643-1GEA 167 EUR
TCIAB1643-1G TCIAB1643-5G
2,2'-Bis(hydroxymethyl)diphenyl ether ≥98.0% (by GC)
2,2'-Bis(hydroxymethyl)diphenyl ether
Formula: C₁₄H₁₄O₃
Peso molecolare: 230,26 g/mol
MDL Number: MFCD00191613
CAS Number: 10038-40-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 100.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR