Benzyl-2-naphthyl ether ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Benzyl 2-Naphthyl Ether, 2-Benzyloxynaphthalene
B1619-25G
B1619-500G
TCIAB1619-25GEA
18
EUR
TCIAB1619-25G
TCIAB1619-500G
Benzyl-2-naphthyl ether ≥98.0% (by GC)
Benzyl-2-naphthyl ether
Formula:
C₁₇H₁₄O Peso molecolare: 234,30 g/mol |
MDL Number:
MFCD00089092 CAS Number: 613-62-7 EINECS: 405-490-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Melting point | 101.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...