Butyraldehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B1594-1G
B1594-10G
TCIAB1594-1GEA
21
EUR
TCIAB1594-1G
TCIAB1594-10G
Butyraldehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Butyraldehyde-2,4-dinitrophenylhydrazone
Formula:
C₁₀H₁₂N₄O₄ Peso molecolare: 252,23 g/mol |
MDL Number:
MFCD00191327 CAS Number: 1527-98-6 UN: 1325 ADR: 4.1,III |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 118.0 to 124.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...