Stampa…

(3-Bromopropyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Bromopropanephosphonic Acid, (3-Bromopropyl)phosphonicacid

B1591-1G B1591-5G
TCIAB1591-1GEA 91 EUR
TCIAB1591-1G TCIAB1591-5G
(3-Bromopropyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(3-Bromopropyl)phosphonic acid
Formula: C₃H₈BrO₃P
Peso molecolare: 202,97 g/mol
MDL Number: MFCD00191385
CAS Number: 1190-09-6
UN: 3261
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR