1,1-Bis(4-hydroxyphenyl)cyclohexane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4'-Cyclohexylidenebisphenol
B1568-25G
B1568-500G
B1568-100G
TCIAB1568-25GEA
25
EUR
TCIAB1568-25G
TCIAB1568-500G
TCIAB1568-100G
1,1-Bis(4-hydroxyphenyl)cyclohexane ≥98.0% (by GC)
1,1-Bis(4-hydroxyphenyl)cyclohexane
Formula:
C₁₈H₂₀O₂ Peso molecolare: 268,36 g/mol |
MDL Number:
MFCD00019341 CAS Number: 843-55-0 EINECS: 212-677-1 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 189.0 to 192.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...