ɑ,ɑ'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4'-(1,4-Phenylenediisopropylidene)bisphenol, 1,4-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene, Bisphenol P
B1563-25G
B1563-500G
TCIAB1563-25GEA
34
EUR
TCIAB1563-25G
TCIAB1563-500G
ɑ,ɑ'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene ≥98.0% (by GC)
alpha,alpha'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene
Formula:
C₂₄H₂₆O₂ Peso molecolare: 346,47 g/mol |
MDL Number:
MFCD00191315 CAS Number: 2167-51-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 194.0 to 197.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...