Stampa…

ɑ,ɑ'-Bis(4-aminophenyl)-1,4-diisopropylbenzene ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,4'-(1,4-Phenylenediisopropylidene)bisaniline, 1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, Bisaniline P

B1562-25G B1562-500G
TCIAB1562-25GEA 40 EUR
TCIAB1562-25G TCIAB1562-500G
ɑ,ɑ'-Bis(4-aminophenyl)-1,4-diisopropylbenzene ≥99.0% (by GC)
alpha,alpha'-Bis(4-aminophenyl)-1,4-diisopropylbenzene
Formula: C₂₄H₂₈N₂
Peso molecolare: 344,50 g/mol
MDL Number: MFCD00191313
CAS Number: 2716-10-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 99.0 %
Melting point 164.0 to 168.0 deg-C
Solubility in hot Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR