Stampa…

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: 2,5-Bis(5-(tert-butyl)benzo[d]oxazol-2-yl)thiophene

B1554-25G B1554-5G
TCIAB1554-25GEA 50 EUR
TCIAB1554-25G TCIAB1554-5G
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥98.0% (by HPLC, total nitrogen)
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
Formula: C₂₆H₂₆N₂O₂S
Peso molecolare: 430,57 g/mol
MDL Number: MFCD00005774
CAS Number: 7128-64-5
EINECS: 230-426-4
Merck Index: 14,01014

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 199.0 to 203.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR