(R)-1-Benzyl-3-pyrrolidinol ≥96.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₁H₁₅NO |
MDL Number:
MFCD00075484 CAS Number: 101930-07-8 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange clear liquid to slightly cloudy liquid |
Purity (GC) | min. 96.0 % |
Specific rotation [a]20/D | -4.4 to -4.8 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...